The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012337.2 | NP_001012337.1 | 1 - 1 | 94 | 102 |
NM_001308313.1 | NP_001295242.1 | 1 - 1 | 94 | 102 | |
XM_005247138.3 | XP_005247195.1 | 1 - 1 | 77 | 102 | |
XM_006713513.3 | XP_006713576.1 | 1 - 1 | 77 | 102 | |
GenBank mRNA | AF231410.1 | - | 1 - 1 | 53 | 102 |
BC141849.1 | - | 1 - 1 | 41 | 102 | |
DC397187.1 | - | 1 - 1 | 94 | 102 | |
HY006578.1 | - | 1 - 1 | 78 | 102 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317774.1 | NP_001304703.1 | 1 - 1 | 86 | 102 |
NM_001317775.1 | NP_001304704.1 | 1 - 1 | 86 | 102 | |
NR_133916.1 | 1 - 1 | 86 | 102 | ||
NR_133918.1 | 1 - 1 | 86 | 102 | ||
NR_133919.1 | 1 - 1 | 86 | 102 | ||
XM_011512933.2 | XP_011511235.1 | 1 - 1 | 84 | 102 | |
XM_011512934.2 | XP_011511236.1 | 1 - 1 | 84 | 102 | |
GenBank mRNA | BC067767.1 | - | 1 - 1 | 53 | 102 |
BG722939.1 | - | 1 - 1 | 46 | 102 | |
BG724071.1 | - | 1 - 1 | 54 | 102 | |
DB042884.1 | - | 1 - 1 | 57 | 102 | |
HY055534.1 | - | 1 - 1 | 86 | 102 |