The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012728.1 | NP_001012746.1 | 3 - 3 | 456 | 89 |
XM_011527011.2 | XP_011525313.1 | 4 - 4 | 516 | 89 | |
XM_011527012.2 | XP_011525314.1 | 4 - 4 | 526 | 89 |