Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- NOTCH2NL
- Entrez Gene ID
-
388677
- Gene Name
- notch 2 N-terminal like
- Gene Aliases
-
N2N
- Chromosome Location
-
Chr.1: 146151908 - 146229032 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_203458.4 |
NP_982283.2 |
5 - 5 |
4158 |
112 |
GenBank mRNA |
BX537434.1 |
- |
6 - 6 |
4226 |
112 |
- Gene Symbol
- LOC100996763
- Entrez Gene ID
-
100996763
- Gene Name
- notch homolog 2 N-terminal-like protein
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 148596477 - 148712497 on Build GRCh38
View Full Product Details