The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032461.3 | NP_115850.2 | 1 - 1 | 154 | 98 |
GenBank mRNA | AF098307.1 | - | 1 - 1 | 154 | 98 |
AJ457796.1 | - | 1 - 1 | 154 | 98 | |
BC034472.1 | - | 1 - 1 | 152 | 98 | |
BC137533.1 | - | 1 - 1 | 144 | 98 | |
BC137536.1 | - | 1 - 1 | 144 | 98 |