Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MIR21
- Entrez Gene ID
-
406991
- Gene Name
- microRNA 21
- Gene Aliases
-
MIRN21, hsa-mir-21, miR-21, miRNA21
- Chromosome Location
-
Chr.17: 59841266 - 59841337 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_029493.1 |
|
1 - 1 |
32 |
64 |
GenBank mRNA |
AF480557.1 |
- |
1 - 1 |
36 |
64 |
AY699265.1 |
- |
1 - 1 |
2477 |
64 |
BC053563.1 |
- |
1 - 1 |
2432 |
64 |
- Gene Symbol
- VMP1
- Entrez Gene ID
-
81671
- Gene Name
- vacuole membrane protein 1
- Gene Aliases
-
EPG3, TANGO5, TMEM49
- Chromosome Location
-
Chr.17: 59707502 - 59840593 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
CB216599.1 |
- |
1 - 1 |
307 |
64 |
View Full Product Details