Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MIR20B
- Entrez Gene ID
-
574032
- Gene Name
- microRNA 20b
- Gene Aliases
-
MIRN20B, hsa-mir-20b, mir-20b
- Chromosome Location
-
Chr.X: 134169809 - 134169877 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_029950.1 |
|
1 - 1 |
33 |
65 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
microRNA 19b-2 |
MIR19B2 |
MIRN19B2, miR-19b-2 |
-43.0
|
Sense |
Chr. - 134169671 - 134169766 on Build GRCh38 |
Downstream |
microRNA 18b |
MIR18B |
MIRN18B, hsa-mir-18b, mir-18b |
164.0
|
Sense |
Chr. - 134170041 - 134170111 on Build GRCh38 |
View Full Product Details