The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_030301.1 | 1 - 1 | 42 | 63 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | long intergenic non-protein coding RNA 575 | LINC00575 | C4orf11 | -131.9kb | Sense | Chr. - 82613113 - 82621437 on Build GRCh38 |
Downstream | SEC31 homolog A, COPII coat complex component | SEC31A | ABP125, ABP130, HSPC275, HSPC334, SEC31L1 | 65.2kb | Sense | Chr. - 82818509 - 82900571 on Build GRCh38 |