Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- RCHY1
- Entrez Gene ID
-
25898
- Gene Name
- ring finger and CHY zinc finger domain containing 1
- Gene Aliases
-
ARNIP, CHIMP, PIRH2, PRO1996, RNF199, ZCHY, ZNF363
- Chromosome Location
-
Chr.4: 75479037 - 75515057 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_037913.1 |
|
7 - 8 |
708 |
100 |
NR_037914.1 |
|
7 - 8 |
704 |
100 |
GenBank mRNA |
AK309555.1 |
- |
7 - 8 |
551 |
100 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC441025 |
LOC441025 |
|
-128.0kb
|
Anti-sense |
Chr. - 75354076 - 75362566 on Build GRCh38 |
Downstream |
THAP domain containing 6 |
THAP6 |
|
18.2kb
|
Anti-sense |
Chr. - 75513951 - 75547334 on Build GRCh38 |
View Full Product Details