Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- SCGB1B2P
- Entrez Gene ID
-
643719
- Gene Name
- secretoglobin family 1B member 2, pseudogene
- Gene Aliases
-
SCGB4A1P
- Chromosome Location
-
Chr.19: 34576733 - 34677031 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_027620.3 |
|
2 - 3 |
1589 |
93 |
GenBank mRNA |
CA429783.1 |
- |
2 - 3 |
|
93 |
CD173361.1 |
- |
1 - 2 |
179 |
93 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
DN831788.1 |
- |
|
|
|
Chr. - - [-] on Build |
93 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
Wilms tumor 1 interacting protein |
WTIP |
|
-68.1kb
|
Anti-sense |
Chr. - 34481935 - 34509147 on Build GRCh38 |
Downstream |
secretoglobin family 2B member 3, pseudogene |
SCGB2B3P |
C2B, SCGB4A3P |
68.1kb
|
Anti-sense |
Chr. - 34645873 - 34646052 on Build GRCh38 |
View Full Product Details