Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- FKBP1AP1
- Entrez Gene ID
-
2282
- Gene Name
- FK506 binding protein 1A pseudogene 1
- Gene Aliases
-
FKBP12, FKBP1P1
- Chromosome Location
-
Chr.19: 57825280 - 57827462 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_024162.1 |
|
2 - 2 |
350 |
92 |
GenBank mRNA |
AI076645.1 |
- |
2 - 2 |
350 |
92 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
H53178.1 |
- |
1 |
|
|
Chr. 19 - 57826434 - 57826817 [+] on Build GRCh38 |
92 |
GenBankmRNA |
R85326.1 |
- |
|
326 |
|
Chr. - - [-] on Build |
92 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
zinc finger protein 552 |
ZNF552 |
|
-10.1kb
|
Sense |
Chr. - 57807082 - 57814913 on Build GRCh38 |
Downstream |
zinc finger protein 587B |
ZNF587B |
|
3.4kb
|
Anti-sense |
Chr. - 57830294 - 57846238 on Build GRCh38 |
View Full Product Details