Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COX16
Entrez Gene ID
51241
Gene Name
COX16, cytochrome c oxidase assembly homolog
Gene Aliases
C14orf112, HSPC203
Chromosome Location
Chr.14: 70325081 - 70359731 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204090.1 NP_001191019.1 1 - 2 215 177
GenBank mRNA CB156527.1 - 1 - 2 159 177

View Full Product Details