The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001195032.1 | NP_001181961.1 | 2 - 3 | 249 | 93 |
NM_001302839.1 | NP_001289768.1 | 2 - 3 | 583 | 93 | |
NM_001302840.1 | NP_001289769.1 | 3 - 4 | 344 | 93 | |
NM_001302841.1 | NP_001289770.1 | 2 - 3 | 243 | 93 | |
XM_005266788.4 | XP_005266845.1 | 2 - 3 | 590 | 93 | |
XM_017010142.1 | XP_016865631.1 | 2 - 3 | 180 | 93 | |
XM_017010143.1 | XP_016865632.1 | 2 - 3 | 257 | 93 | |
XM_017010144.1 | XP_016865633.1 | 2 - 3 | 252 | 93 | |
XM_017010145.1 | XP_016865634.1 | 1 - 2 | 155 | 93 | |
XM_017010146.1 | XP_016865635.1 | 1 - 2 | 150 | 93 | |
XM_017010147.1 | XP_016865636.1 | 2 - 3 | 240 | 93 | |
GenBank mRNA | AW138390.1 | - | 1 - 2 | 93 | |
BF688381.1 | - | 2 - 3 | 250 | 93 | |
HY052980.1 | - | 3 - 4 | 345 | 93 | |
HY205185.1 | - | 1 - 2 | 93 | ||
W37337.1 | - | 2 - 3 | 171 | 93 |