Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WISP1
Entrez Gene ID
8840
Gene Name
WNT1 inducible signaling pathway protein 1
Gene Aliases
CCN4, WISP1c, WISP1i, WISP1tc
Chromosome Location
Chr.8: 133191039 - 133231690 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204869.1 NP_001191798.1 1 - 2 182 84
NM_003882.3 NP_003873.1 1 - 2 182 84
NM_080838.2 NP_543028.1 1 - 2 182 84
GenBank mRNA AK293031.1 - 1 - 2 182 84
AK315822.1 - 1 - 2 182 84
AY196488.1 - 1 - 2 137 84
BC074840.2 - 1 - 2 105 84
BC074841.2 - 1 - 2 105 84
BX476382.1 - 1 - 2 182 84
DN997366.1 - 1 - 2 169 84

View Full Product Details