The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004617.3 | NP_004608.1 | 1 - 2 | 464 | 98 |
GenBank mRNA | AK055577.1 | - | 1 - 2 | 1071 | 98 |
AK315464.1 | - | 1 - 2 | 351 | 98 | |
BC001386.2 | - | 1 - 2 | 334 | 98 | |
BP294159.1 | - | 1 - 2 | 464 | 98 | |
BT007430.1 | - | 1 - 2 | 167 | 98 | |
U31449.1 | - | 1 - 2 | 332 | 98 |