The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006418.4 | NP_006409.3 | 1 - 2 | 299 | 159 |
GenBank mRNA | AF097021.1 | - | 1 - 2 | 217 | 159 |
AK300947.1 | - | 1 - 2 | 179 | 159 | |
AY358567.1 | - | 1 - 2 | 216 | 159 | |
BC117329.1 | - | 1 - 2 | 281 | 159 | |
DA928098.1 | - | 1 - 2 | 304 | 159 |