Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CDRT7
- Entrez Gene ID
-
94150
- Gene Name
- CMT1A duplicated region transcript 7 (non-protein coding)
- Gene Aliases
-
LINC00025, NCRNA00025
- Chromosome Location
-
Chr.17: 15030975 - 15031957 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_033371.1 |
|
2 - 3 |
398 |
90 |
GenBank mRNA |
AI218510.1 |
- |
1 - 2 |
|
90 |
BX093265.1 |
- |
2 - 3 |
398 |
90 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC101928475 |
LOC101928475 |
|
|
Sense |
Chr. - 15051355 - 15052276 on Build GRCh38 |
Downstream |
CMT1A duplicated region transcript 8 |
CDRT8 |
|
73.0kb
|
Anti-sense |
Chr. - 15104985 - 15106187 on Build GRCh38 |
View Full Product Details