The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_033760.1 | 1 - 1 | 4221 | 138 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | transmembrane protein 14A | TMEM14A | C6orf73, PTD011 | -52.9kb | Anti-sense | Chr. - 52671086 - 52686588 on Build GRCh38 |
Downstream | glutathione S-transferase alpha 2 | GSTA2 | GST2, GSTA2-2, GTA2, GTH2 | 4.9kb | Sense | Chr. - 52750087 - 52763563 on Build GRCh38 |