Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- SNORA58
- Entrez Gene ID
-
677836
- Gene Name
- small nucleolar RNA, H/ACA box 58
- Gene Aliases
-
ACA58, SNORA58B
- Chromosome Location
-
Chr.3: 131478998 - 131479233 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_002985.2 |
|
1 - 1 |
121 |
120 |
GenBank mRNA |
DW436402.1 |
- |
1 - 1 |
137 |
120 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
DW451474.1 |
- |
1 |
137 |
|
Chr. 3 - 131478873 - 131479236 [-] on Build GRCh38 |
120 |
GenBankmRNA |
DW421138.1 |
- |
1 |
137 |
|
Chr. 3 - 131478953 - 131479236 [-] on Build GRCh38 |
120 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
nudix hydrolase 16 |
NUDT16 |
|
-90.2kb
|
Anti-sense |
Chr. - 131381671 - 131388830 on Build GRCh38 |
Downstream |
BCL2 like 12 pseudogene 1 |
BCL2L12P1 |
|
47.2kb
|
Anti-sense |
Chr. - 131526468 - 131527141 on Build GRCh38 |
View Full Product Details