Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BTG4
Entrez Gene ID
54766
Gene Name
BTG anti-proliferation factor 4
Gene Aliases
APRO3, PC3B
Chromosome Location
Chr.11: 111383828 - 111512388 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017589.3 NP_060059.1 2 - 3 373 106
XM_011542876.2 XP_011541178.1 2 - 3 407 106
XM_011542878.2 XP_011541180.1 2 - 3 407 106
XM_011542879.2 XP_011541181.1 2 - 3 407 106
XM_011542880.2 XP_011541182.1 2 - 3 407 106
GenBank mRNA AJ271351.1 - 1 - 2 178 106
BC031045.1 - 2 - 3 359 106
BX099301.1 - 2 - 3 373 106

View Full Product Details