Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
SLC29A1
Entrez Gene ID
2030
Gene Name
solute carrier family 29 member 1 (Augustine blood group)
Gene Aliases
ENT1
Chromosome Location
Chr.6: 44219505 - 44234151 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001304466.1 NP_001291395.1 2 - 2 296 80
GenBank mRNA AX775997.1 - 2 - 2 145 80
U81375.1 - 2 - 2 145 80

View Full Product Details