Product Details

Assay Reference Genome
Location

Chr.7:100420325 on build GRCh38
Cytoband
7q22.1
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
55063
Gene Symbol
ZCWPW1
Gene Name
zinc finger CW-type and PWWP domain containing 1
Gene Aliases
ZCW1
Location
Chr.7:100400826-100429239 on build GRCh38
Assay Gene Location
Within Intron 4
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
ZCWPW1 NM_001258008.1 NP_001244937.1
NM_017984.4 NP_060454.3
XM_005250479.2 XP_005250536.1
XM_005250480.2 XP_005250537.1
XM_006716035.3 XP_006716098.1
XM_006716036.2 XP_006716099.1
XM_006716037.3 XP_006716100.1
XM_006716038.3 XP_006716101.1
XM_006716040.3 XP_006716103.1
XM_017012379.1 XP_016867868.1
XM_017012380.1 XP_016867869.1
AK093038.1 BAC04028.1
AK299989.1
AK300745.1
AK302108.1
AK316553.1
AL136735.1 CAB66669.1
DB447154.1
DC413605.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv2734888 Chr7:99975623 - 101290285 on Build GRCh38 Deletion NYAP1 AP1S1 ZNF3 COPS6 SAP25 VGF MIR4653 ZASP MIR4658 NAT16 PVRIG PILRB PILRA POP7 PLOD3 TFR2 TAF6 MUC17 EPHB4 MIR93 CNPY4 MIR25 CLDN15 AZGP1 LRCH4 SPDYE3 C7orf43 GAL3ST4 PVRIG2P STAG3 MEPCE MOGAT3 MUC3A STAG3L5P MCM7 ZAN MIR6840 ZCWPW1 GNB2 TRIP6 ZKSCAN1 ZNHIT1 FBXO24 ACHE SLC12A9 MUC12 ZSCAN21 MIR106B SERPINE1 AGFG2 C7orf61 MBLAC1 LOC105375429 AZGP1P1 ACTL6B STAG3L5P-PVRIG2P-PILRB GATS GIGYF1 MIR6875 FIS1 SRRT AP4M1 PPP1R35 PCOLCE-AS1 UFSP1 LOC102724094 PCOLCE PMS2P1 TRIM56 GPC2 TSC22D4 EPO LAMTOR4 MOSPD3
nsv831071 Chr7:100320402 - 100515790 on Build GRCh38 Loss NYAP1 SPDYE3 PPP1R35 C7orf61 PVRIG2P MEPCE STAG3L5P PILRB PILRA PMS2P1 MIR6840 STAG3L5P-PVRIG2P-PILRB ZCWPW1 TSC22D4
nsv527107 Chr7:100417088 - 100467096 on Build GRCh38 Loss MEPCE ZCWPW1 TSC22D4 PPP1R35 C7orf61

View Full Product Details