Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GAS6-AS1
Entrez Gene ID
650669
Gene Name
GAS6 antisense RNA 1
Gene Aliases
-
Chromosome Location
Chr.13: 113815610 - 113842841 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_044995.2 3 - 4 251 110
GenBank mRNA DA460725.1 - 3 - 4 249 110

View Full Product Details