Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- PRRT3-AS1
- Entrez Gene ID
-
100874032
- Gene Name
- PRRT3 antisense RNA 1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.3: 9947404 - 9954787 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_046734.1 |
|
1 - 2 |
196 |
177 |
GenBank mRNA |
AI861893.1 |
- |
3 - 4 |
|
177 |
BQ000843.1 |
- |
3 - 4 |
|
177 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
cysteine rich with EGF like domains 1 |
CRELD1 |
AVSD2, CIRRIN |
-2.0kb
|
Sense |
Chr. - 9933788 - 9945413 on Build GRCh38 |
Downstream |
ER membrane protein complex subunit 3 |
EMC3 |
POB, TMEM111 |
14.5kb
|
Anti-sense |
Chr. - 9962680 - 10011248 on Build GRCh38 |
View Full Product Details