The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270629.1 | NP_001257558.1 | 4 - 5 | 376 | 108 |
NM_014166.3 | NP_054885.1 | 4 - 5 | 482 | 108 | |
XM_017020549.1 | XP_016876038.1 | 4 - 5 | 1570 | 108 | |
GenBank mRNA | AF161475.1 | - | 4 - 5 | 444 | 108 |
AF230381.1 | - | 4 - 5 | 419 | 108 | |
AF300618.1 | - | 4 - 5 | 419 | 108 | |
AK001934.1 | - | 4 - 5 | 445 | 108 | |
AK222713.1 | - | 4 - 5 | 445 | 108 | |
AK301835.1 | - | 4 - 5 | 350 | 108 | |
BC005189.1 | - | 4 - 5 | 456 | 108 | |
CR457276.1 | - | 4 - 5 | 419 | 108 | |
CR541830.1 | - | 4 - 5 | 419 | 108 | |
HG518167.1 | - | 4 - 5 | 482 | 108 |