Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CAMKMT
Entrez Gene ID
79823
Gene Name
calmodulin-lysine N-methyltransferase
Gene Aliases
C2orf34, CLNMT, CaM KMT, Cam, KMT
Chromosome Location
Chr.2: 44361714 - 44774388 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024766.4 NP_079042.1 8 - 9 802 75
XM_011533113.2 XP_011531415.1 6 - 7 478 75
XM_017004982.1 XP_016860471.1 6 - 7 2905 75
XM_017004983.1 XP_016860472.1 7 - 8 1017 75
GenBank mRNA AK027104.1 - 8 - 9 742 75
BC053733.1 - 8 - 9 729 75

View Full Product Details