The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001253379.1 | NP_001240308.1 | 2 - 3 | 568 | 87 |
NM_001253380.1 | NP_001240309.1 | 1 - 2 | 330 | 87 | |
NM_001253382.1 | NP_001240311.1 | 2 - 3 | 319 | 87 | |
NM_001253383.1 | NP_001240312.1 | 2 - 3 | 298 | 87 | |
NM_001253384.1 | NP_001240313.1 | 2 - 3 | 555 | 87 | |
NM_002948.3 | NP_002939.2 | 2 - 3 | 555 | 87 | |
GenBank mRNA | AF279903.1 | - | 1 - 2 | 209 | 87 |
AF283772.2 | - | 2 - 3 | 215 | 87 | |
AK098795.1 | - | 2 - 3 | 205 | 87 | |
AK129815.1 | - | 2 - 3 | 215 | 87 | |
AK290694.1 | - | 2 - 3 | 211 | 87 | |
AK292000.1 | - | 2 - 3 | 211 | 87 | |
AK293709.1 | - | 2 - 3 | 211 | 87 | |
AK297091.1 | - | 2 - 3 | 213 | 87 | |
AY347528.1 | - | 1 - 2 | 280 | 87 | |
BC014837.1 | - | 2 - 3 | 218 | 87 | |
BC068198.1 | - | 1 - 1 | 214 | 87 | |
BC070328.1 | - | 2 - 3 | 211 | 87 | |
BC071672.1 | - | 1 - 1 | 200 | 87 | |
BC081565.1 | - | 2 - 3 | 220 | 87 | |
BE893727.1 | - | 2 - 3 | 290 | 87 | |
BT019672.1 | - | 1 - 2 | 175 | 87 | |
BU178524.1 | - | 2 - 3 | 319 | 87 | |
DA037869.1 | - | 2 - 3 | 236 | 87 | |
DB461802.1 | - | 1 - 2 | 330 | 87 | |
DC351786.1 | - | - | 298 | 87 | |
L25899.1 | - | 1 - 2 | 191 | 87 |