The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270766.1 | NP_001257695.1 | 8 - 9 | 749 | 73 |
NM_001270767.1 | NP_001257696.1 | 7 - 8 | 683 | 73 | |
NM_173474.3 | NP_775745.1 | 9 - 10 | 852 | 73 | |
XM_011522355.2 | XP_011520657.1 | 6 - 7 | 687 | 73 | |
GenBank mRNA | AK092915.1 | - | 5 - 6 | 2035 | 73 |
BC017336.2 | - | 9 - 10 | 749 | 73 |