Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HYDIN
Entrez Gene ID
54768
Gene Name
HYDIN, axonemal central pair apparatus protein
Gene Aliases
CILD5, HYDIN1, HYDIN2, PPP1R31
Chromosome Location
Chr.16: 70802084 - 71230722 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001270974.2 NP_001257903.1 73 - 74 12623 75
XM_006721206.2 XP_006721269.1 73 - 74 12517 75
XM_011523146.2 XP_011521448.1 74 - 75 12652 75
XM_011523147.1 XP_011521449.1 74 - 75 12619 75
XM_011523148.1 XP_011521450.1 74 - 75 12692 75
XM_011523151.1 XP_011521453.1 73 - 74 12549 75
XM_011523152.1 XP_011521454.1 36 - 37 6376 75
XM_011523155.2 XP_011521457.1 29 - 30 5377 75
XM_017023346.1 XP_016878835.1 73 - 74 12795 75
XM_017023347.1 XP_016878836.1 61 - 62 11034 75
XM_017023348.1 XP_016878837.1 61 - 62 10774 75
GenBank mRNA BC043273.2 - 8 - 9 1343 75

View Full Product Details