The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_109962.1 | 1 - 2 | 56 | 100 | |
GenBank mRNA | BC034606.1 | - | 1 - 2 | 56 | 100 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC400867 | LOC400867 | -39.0kb | Sense | Chr. - 38877291 - 38938429 on Build GRCh38 | |
Downstream | proteasome assembly chaperone 1 | PSMG1 | C21LRP, DSCR2, LRPC21, PAC-1, PAC1 | 197.7kb | Sense | Chr. - 39175446 - 39183514 on Build GRCh38 |