Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC101928435
Entrez Gene ID
101928435
Gene Name
uncharacterized LOC101928435
Gene Aliases
-
Chromosome Location
Chr.21: 38974429 - 38977774 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_109962.1 2 - 3 181 64
GenBank mRNA BC034606.1 - 2 - 3 181 64


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BG721537.1 - 2 - 3 186 Chr. 21 - 38975286 - 38977774 [-] on Build GRCh38 64


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream uncharacterized LOC400867 LOC400867 -36.0kb Sense Chr. - 38877291 - 38938429 on Build GRCh38
Downstream proteasome assembly chaperone 1 PSMG1 C21LRP, DSCR2, LRPC21, PAC-1, PAC1 197.9kb Sense Chr. - 39175446 - 39183514 on Build GRCh38

View Full Product Details