Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BU678341.1 - 1 475 Chr. 4 - 119500048 - 119500610 [-] on Build GRCh38 103


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream septin 7 pseudogene LOC645513 -456.0 Anti-sense Chr. - 119454783 - 119499592 on Build GRCh38
Downstream long intergenic non-protein coding RNA 1365 LINC01365 298.5kb Sense Chr. - 119799106 - 119804515 on Build GRCh38

View Full Product Details