The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001300812.1 | NP_001287741.1 | 2 - 3 | 353 | 136 |
GenBank mRNA | BC144645.1 | - | 2 - 3 | 353 | 136 |