The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001300815.1 | NP_001287744.1 | 6 - 7 | 650 | 91 |
XM_017026237.1 | XP_016881726.1 | 6 - 7 | 711 | 91 | |
GenBank mRNA | BM706055.1 | - | 6 - 7 | 91 | |
BU689520.1 | - | 1 - 2 | 91 |