The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001082969.1 | NP_001076438.1 | 4 - 5 | 449 | 143 |
NM_001301019.1 | NP_001287948.1 | 4 - 5 | 490 | 143 | |
NM_018195.3 | NP_060665.3 | 4 - 5 | 1007 | 143 | |
NR_103469.1 | 4 - 5 | 1048 | 143 | ||
GenBank mRNA | AB096251.1 | - | 4 - 5 | 353 | 143 |
BC005403.1 | - | 2 - 3 | 246 | 143 | |
BC030546.1 | - | 2 - 3 | 251 | 143 | |
BC048133.1 | - | 4 - 5 | 935 | 143 | |
BG115155.1 | - | 4 - 5 | 497 | 143 | |
BX538107.1 | - | 4 - 5 | 1006 | 143 | |
CD244552.1 | - | 4 - 5 | 408 | 143 |