The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001082969.1 | NP_001076438.1 | 5 - 6 | 496 | 93 |
NM_001301019.1 | NP_001287948.1 | 5 - 6 | 537 | 93 | |
NM_018195.3 | NP_060665.3 | 5 - 6 | 1054 | 93 | |
NR_103469.1 | 5 - 6 | 1095 | 93 | ||
GenBank mRNA | AB096251.1 | - | 5 - 6 | 400 | 93 |
AK001588.1 | - | 2 - 3 | 112 | 93 | |
BC005403.1 | - | 3 - 4 | 293 | 93 | |
BC030546.1 | - | 3 - 4 | 298 | 93 | |
BC048133.1 | - | 5 - 6 | 982 | 93 | |
BX538107.1 | - | 5 - 6 | 1053 | 93 | |
CD244552.1 | - | 5 - 6 | 455 | 93 |