Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CCDC53
Entrez Gene ID
51019
Gene Name
coiled-coil domain containing 53
Gene Aliases
CGI-116
Chromosome Location
Chr.12: 102012840 - 102062124 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001301107.1 NP_001288036.1 4 - 5 481 63
GenBank mRNA BX356987.2 - 4 - 5 436 63

View Full Product Details