The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301873.1 | NP_001288802.1 | 2 - 3 | 268 | 167 |
NM_001301874.1 | NP_001288803.1 | 2 - 3 | 268 | 167 | |
NM_001301875.1 | NP_001288804.1 | 2 - 3 | 359 | 167 | |
NM_148672.3 | NP_683513.1 | 2 - 3 | 268 | 167 | |
GenBank mRNA | AF110384.1 | - | 2 - 3 | 247 | 167 |
AF220210.1 | - | 2 - 3 | 193 | 167 | |
AF266504.1 | - | 2 - 3 | 245 | 167 | |
AK307307.1 | - | 2 - 3 | 223 | 167 | |
AK312120.1 | - | 2 - 3 | 251 | 167 | |
BC062668.1 | - | 2 - 3 | 210 | 167 | |
BC069532.1 | - | 2 - 3 | 243 | 167 | |
BX486632.1 | - | 2 - 3 | 269 | 167 | |
CD671712.1 | - | 2 - 3 | 359 | 167 |