Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- NANOG
- Entrez Gene ID
-
79923
- Gene Name
- Nanog homeobox
- Gene Aliases
-
-
- Chromosome Location
-
Chr.12: 7789396 - 7796061 on Build GRCh38
- Gene Symbol
- NANOGP1
- Entrez Gene ID
-
404635
- Gene Name
- Nanog homeobox pseudogene 1
- Gene Aliases
-
NANOG2
- Chromosome Location
-
Chr.12: 7892356 - 7899246 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
KC713782.1 |
- |
1 - 2 |
157 |
101 |
View Full Product Details