Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MED12L
Entrez Gene ID
116931
Gene Name
mediator complex subunit 12 like
Gene Aliases
NOPAR, TNRC11L, TRALP, TRALPUSH
Chromosome Location
Chr.3: 151085665 - 151436677 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053002.5 NP_443728.3 2 - 3 329 103
XM_006713487.3 XP_006713550.1 3 - 4 601 103
XM_011512390.2 XP_011510692.1 3 - 4 601 103
XM_011512394.2 XP_011510696.1 3 - 4 601 103
XM_011512399.2 XP_011510701.1 3 - 4 601 103
XM_017005676.1 XP_016861165.1 3 - 4 601 103
XM_017005677.1 XP_016861166.1 3 - 4 601 103
XM_017005678.1 XP_016861167.1 3 - 4 601 103
XM_017005679.1 XP_016861168.1 3 - 4 601 103
XM_017005680.1 XP_016861169.1 3 - 4 601 103
GenBank mRNA AF388364.1 - 2 - 3 733 103
AF388365.1 - 2 - 3 733 103
AF399708.1 - 2 - 3 238 103
AK128165.1 - 3 - 4 602 103
BC144559.1 - 2 - 3 233 103

View Full Product Details