Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ART5
Entrez Gene ID
116969
Gene Name
ADP-ribosyltransferase 5
Gene Aliases
ARTC5
Chromosome Location
Chr.11: 3638503 - 3642392 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079536.1 NP_001073004.1 2 - 3 197 103
NM_001297668.1 NP_001284597.1 1 - 2 510 103
NM_053017.4 NP_443750.2 1 - 2 510 103
XM_005252779.3 XP_005252836.1 1 - 2 186 103
XM_006718136.3 XP_006718199.1 2 - 3 81 103
XM_011519879.2 XP_011518181.1 2 - 3 231 103
GenBank mRNA AK313531.1 - 1 - 2 186 106
AY358466.1 - 1 - 2 132 106
BC014577.1 - 1 - 2 198 106
CD557970.1 - 1 - 2 449 106
Y16835.1 - 2 - 3 197 106

View Full Product Details