Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HTRA3
Entrez Gene ID
94031
Gene Name
HtrA serine peptidase 3
Gene Aliases
Prsp, Tasp
Chromosome Location
Chr.4: 8269757 - 8307111 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001297559.1 NP_001284488.1 2 - 3 690 97
NM_053044.4 NP_444272.1 2 - 3 690 97
XM_011513596.2 XP_011511898.1 2 - 3 698 97
GenBank mRNA AL547377.3 - 2 - 3 693 97
AY040094.2 - 2 - 3 707 97
AY280665.1 - 2 - 3 678 97
AY280666.1 - 2 - 3 593 97
BC034390.1 - 2 - 3 690 97
BC035717.1 - 2 - 3 690 97

View Full Product Details