Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FCRL2
Entrez Gene ID
79368
Gene Name
Fc receptor like 2
Gene Aliases
CD307b, FCRH2, IFGP4, IRTA4, SPAP1, SPAP1A, SPAP1B, SPAP1C
Chromosome Location
Chr.1: 157745733 - 157777472 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159488.1 NP_001152960.1 8 - 9 1364 98

View Full Product Details