The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278645.1 | NP_001265574.1 | 2 - 3 | 345 | 67 |
NM_001278646.1 | NP_001265575.1 | 2 - 3 | 378 | 67 | |
NM_001311168.1 | NP_001298097.1 | 2 - 3 | 347 | 67 | |
NM_005005.2 | NP_004996.1 | 2 - 3 | 380 | 67 | |
GenBank mRNA | AF044956.1 | - | 2 - 3 | 374 | 67 |
AF067168.1 | - | 2 - 3 | 374 | 67 | |
AF261090.1 | - | 2 - 3 | 379 | 67 | |
AK026538.1 | - | 2 - 3 | 363 | 67 | |
AK302286.1 | - | 2 - 3 | 342 | 67 | |
AK313432.1 | - | 2 - 3 | 342 | 67 | |
AL136714.1 | - | 2 - 3 | 350 | 67 | |
BC007672.2 | - | 2 - 3 | 347 | 67 | |
BG703246.1 | - | 2 - 3 | 307 | 67 | |
BQ685604.1 | - | 2 - 3 | 330 | 67 | |
CN410734.1 | - | 2 - 3 | 288 | 67 | |
DB504123.1 | - | 2 - 3 | 305 | 67 | |
S82655.1 | - | 1 - 2 | 66 | 67 |