The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282276.1 | NP_001269205.1 | 1 - 2 | 82 | 110 |
GenBank mRNA | BX106051.1 | - | 1 - 2 | 82 | 110 |