Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CNTLN
Entrez Gene ID
54875
Gene Name
centlein
Gene Aliases
C9orf101, C9orf39, bA340N12.1
Chromosome Location
Chr.9: 17134991 - 17510019 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286984.1 NP_001273913.1 2 - 3 530 119
NM_001286985.1 NP_001273914.1 2 - 3 341 119
GenBank mRNA AK303973.1 - 2 - 3 488 119
BU663791.1 - 2 - 3 333 119
DA482759.1 - 2 - 3 482 119

View Full Product Details