The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001294139.1 | NP_001281068.1 | 2 - 2 | 1226 | 98 |
NM_023013.4 | NP_075389.2 | 4 - 4 | 1916 | 98 | |
GenBank mRNA | AK125568.1 | - | 5 - 5 | 1889 | 98 |
BC040902.1 | - | 2 - 2 | 1226 | 98 |