Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- ZXDC
- Entrez Gene ID
-
79364
- Gene Name
- ZXD family zinc finger C
- Gene Aliases
-
ZXDL
- Chromosome Location
-
Chr.3: 126437348 - 126475938 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_104249.1 |
|
6 - 7 |
2452 |
96 |
GenBank mRNA |
BC063713.1 |
- |
6 - 7 |
1520 |
96 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
cilia and flagella associated protein 100 |
CFAP100 |
CCDC37, MIA1 |
-5.2kb
|
Anti-sense |
Chr. - 126394893 - 126436556 on Build GRCh38 |
Downstream |
urocanate hydratase 1 |
UROC1 |
HMFN0320, UROCD |
18.9kb
|
Sense |
Chr. - 126481165 - 126517773 on Build GRCh38 |
View Full Product Details