Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- NEDD4
- Entrez Gene ID
-
4734
- Gene Name
- neural precursor cell expressed, developmentally down-regulated 4, E3 ubiquitin protein ligase
- Gene Aliases
-
NEDD4-1, RPF1
- Chromosome Location
-
Chr.15: 55826917 - 55993746 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_104302.1 |
|
1 - 2 |
1850 |
112 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
protogenin |
PRTG |
IGDCC5 |
-125.9kb
|
Sense |
Chr. - 55611540 - 55743649 on Build GRCh38 |
Downstream |
regulatory factor X7 |
RFX7 |
RFXDC2 |
173.4kb
|
Sense |
Chr. - 56090533 - 56247654 on Build GRCh38 |
View Full Product Details