Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MIR6069
- Entrez Gene ID
-
102464824
- Gene Name
- microRNA 6069
- Gene Aliases
-
hsa-mir-6069
- Chromosome Location
-
Chr.22: 35336721 - 35336799 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_106717.1 |
|
1 - 1 |
38 |
65 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
microRNA 3909 |
MIR3909 |
mir-3909 |
-963.0
|
Anti-sense |
Chr. - 35335640 - 35335758 on Build GRCh38 |
Downstream |
heme oxygenase 1 |
HMOX1 |
HMOX1D, HO-1, HSP32, bK286B10 |
44.3kb
|
Anti-sense |
Chr. - 35381067 - 35394214 on Build GRCh38 |
View Full Product Details