Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- LINC01506
- Entrez Gene ID
-
101927015
- Gene Name
- long intergenic non-protein coding RNA 1506
- Gene Aliases
-
-
- Chromosome Location
-
Chr.9: 68543541 - 68546602 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_109769.1 |
|
3 - 4 |
290 |
116 |
GenBank mRNA |
CR745134.1 |
- |
3 - 4 |
290 |
116 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AI271718.1 |
- |
1 - 2 |
|
|
Chr. 9 - 68543542 - 68545838 [+] on Build GRCh38 |
116 |
GenBankmRNA |
AW451215.1 |
- |
1 - 2 |
|
|
Chr. 9 - 68543541 - 68546589 [+] on Build GRCh38 |
116 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC105376072 |
LOC105376072 |
|
|
Anti-sense |
Chr. - 68541154 - 68644340 on Build GRCh38 |
Downstream |
phosphatidylinositol-4-phosphate 5-kinase type 1 beta |
PIP5K1B |
MSS4, STM7 |
159.6kb
|
Anti-sense |
Chr. - 68705217 - 69009176 on Build GRCh38 |
View Full Product Details